trans-Crotononitrile 75,0 GC_ASSAY_METHOD (contains ca. 20% cis- isomer)

Supplier: TCI America
Danger

Synonyms: trans-2-Butenenitrile

C3132-100ML C3132-25ML
TCC3132-100MLEA 285.98 CAD
TCC3132-100ML TCC3132-25ML
trans-Crotononitrile 75,0 GC_ASSAY_METHOD (contains ca. 20% cis- isomer)
trans-Crotononitrile
Formula: C₄H₅N
MW: 67.09 g/mol
Flash Pt: 16°C
CAS Number: 627-26-9
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 75.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR