Potassium trimethoxy(trifluoromethyl)borate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

P2036-25G P2036-5G
TCP2036-25GEA 801.06 CAD
TCP2036-25G TCP2036-5G
Potassium trimethoxy(trifluoromethyl)borate ≥95.0% (by titrimetric analysis)
Potassium trimethoxy(trifluoromethyl)borate
Formula: C₄H₉BF₃KO₃
Melting Pt: 118°C
Storage Temperature: Refrigerator
MDL Number: MFCD22124929
CAS Number: 626232-27-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR