1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B4844-25G B4844-5G
TCB4844-25GEA 330.54 CAD
TCB4844-25G TCB4844-5G
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide
Formula: C₁₂H₂₂F₆N₂O₄S₂
MDL Number: MFCD14156017
CAS Number: 623580-02-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR