2-Hexyldecan-1-amine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
H1634-1ML
H1634-5ML
TCH1634-1MLEA
281.24
CAD
TCH1634-1ML
TCH1634-5ML
2-Hexyldecan-1-amine ≥98.0% (by GC, titration analysis)
2-Hexyldecan-1-amine
Formula:
C₁₆H₃₅N Boiling Pt: 150°C |
MDL Number:
MFCD30475384 CAS Number: 62281-05-4 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...