Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Sodium 5-Isopropyl-3,8-dimethylazulene-1-sulphonate, Sodium Guaiazulenesulfonate, Guaiazulenesulfonic acid sodium salt, Sodium Guaiazulenesulphonate, 7-Isopropyl-1,4-dimethyl-azulene-3-sulfonic acid sodium salt, Sodium 5-Isopropyl-3,8-dimethylazulene-1-sulfonate, 7-Isopropyl-1,4-dimethyl-azulene-3-sulphonic acid sodium salt, Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate, 5-Isopropyl-3,8-dimethylazulene-1-sulfonic acid sodium salt, 5-Isopropyl-3,8-dimethylazulene-1-sulphonic acid sodium salt, Guaiazulenesulphonic acid sodium salt, Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate

S0976-1G S0976-5G
TCS0976-1GEA 67.94 CAD
TCS0976-1G TCS0976-5G
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate ≥98.0% (by HPLC, titration analysis)
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate
Formula: C₁₅H₁₇NaO₃S
MW: 300.35 g/mol
Melting Pt: 98°C
Storage Temperature: Freezer
MDL Number: MFCD00866167
CAS Number: 6223-35-4
EINECS: 228-309-8
Merck Index: 14,04554

Order Now

Specification Test Results

Appearance Blue to Dark blue powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 2.5 to 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR