2-Bromophenanthrene ≥97.0% (by GC)

Supplier: TCI America

B4671-1G B4671-200MG
TCB4671-1GEA 233.84 CAD
TCB4671-1G TCB4671-200MG
2-Bromophenanthrene ≥97.0% (by GC)
2-Bromophenanthrene
Formula: C₁₄H₉Br
Boiling Pt: 185°C
MDL Number: MFCD13191756
CAS Number: 62162-97-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR