2,7-Dimethoxy-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5329-1G D5329-5G
76219-912EA 214.88 CAD
76219-912 76219-914
2,7-Dimethoxy-9H-carbazole ≥98.0% (by GC)
2,7-Dimethoxy-9H-carbazole
Formula: C₁₄H₁₃NO₂
MDL Number: MFCD28154716
CAS Number: 61822-18-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR