1,5-Dibromo-2,4-dimethylbenzene ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
D5004-25G
D5004-5G
76219-120EA
665.18
CAD
76219-120
76219-122
1,5-Dibromo-2,4-dimethylbenzene ≥97.0% (by GC)
1,5-Dibromo-2,4-dimethylbenzene
Formula:
C₈H₈Br₂ Boiling Pt: 256°C |
MDL Number:
MFCD00154984 CAS Number: 615-87-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...