1-Methyl-2-(methylsulfonyl)benzimidazole ≥98.0% (by GC)
Supplier: TCI America
M2860-1G
M2860-5G
76220-288EA
227.52
CAD
76220-288
76220-290
1-Methyl-2-(methylsulfonyl)benzimidazole ≥98.0% (by GC)
1-Methyl-2-(methylsulfonyl)benzimidazole
Formula:
C₉H₁₀N₂O₂S |
CAS Number:
61078-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...