1,2,3-Tribromobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

T3329-1G T3329-5G
76220-562EA 213.3 CAD
76220-562 76220-564
1,2,3-Tribromobenzene ≥98.0% (by GC)
1,2,3-Tribromobenzene
Formula: C₆H₃Br₃
Boiling Pt: 283°C
MDL Number: MFCD00192668
CAS Number: 608-21-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR