Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3208-1G T3208-200MG
TCT3208-1GEA 375.25 CAD
TCT3208-1G TCT3208-200MG
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) ≥98.0% (by HPLC)
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate)
Formula: C₃₆H₂₄F₁₂N₆P₂Ru
MDL Number: MFCD28369263
CAS Number: 60804-75-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.50 to 9.50 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR