2-Methylbenzoic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2997-1G M2997-5G
TCM2997-1GEA 192.76 CAD
TCM2997-1G TCM2997-5G
2-Methylbenzoic anhydride ≥98.0% (by GC, titration analysis)
2-Methylbenzoic anhydride
Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Boiling Pt: 221°C
MDL Number: MFCD00048077
CAS Number: 607-86-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR