3-Ethoxy-4-methoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

E1109-25G E1109-5G
TCE1109-25GEA 308.1 CAD
TCE1109-25G TCE1109-5G
3-Ethoxy-4-methoxybenzonitrile ≥98.0% (by GC)
3-Ethoxy-4-methoxybenzonitrile
Formula: C₁₀H₁₁NO₂
MDL Number: MFCD02256160
CAS Number: 60758-86-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR