3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
C3110-1G
C3110-5G
TCC3110-1GEA
116.92
CAD
TCC3110-1G
TCC3110-5G
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride
Formula:
C₁₀H₈O₆ |
MDL Number:
MFCD22373052 CAS Number: 6053-46-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Methanolysis Method) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...