11,12-Dihydroindolo[2,3-a]carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
D4803-1G
D4803-200MG
TCD4803-1GEA
158
CAD
TCD4803-1G
TCD4803-200MG
11,12-Dihydroindolo[2,3-a]carbazole ≥98.0% (by HPLC, total nitrogen)
11,12-Dihydroindolo[2,3-a]carbazole
Formula:
C₁₈H₁₂N₂ Melting Pt: 371°C |
MDL Number:
MFCD07371389 CAS Number: 60511-85-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...