3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Tris[dimethyl(vinyl)siloxy]phenylsilane
D5264-25G
D5264-5G
TCD5264-25GEA
412.38
CAD
TCD5264-25G
TCD5264-5G
3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane ≥98.0% (by GC)
3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane
Formula:
C₁₈H₃₂O₃Si₄ MW: 408.79 g/mol |
MDL Number:
MFCD00156495 CAS Number: 60111-47-9 EINECS: 262-056-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...