2,3-Thiophenedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Thieno[2,3-c]furan-4,6-dione
T3342-1G
T3342-5G
76220-570EA
214.88
CAD
76220-570
76220-572
2,3-Thiophenedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
2,3-Thiophenedicarboxylic anhydride
Formula:
C₆H₂O₃S MW: 154.14 g/mol |
MDL Number:
MFCD09832453 CAS Number: 6007-83-6 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...