VWR® Flow-Through and Immersion Coolers
Supplier: VWR International
13271-500 13271-502 13271-504 89400-956 89400-960
13271-500EA 6001.08 CAD
13271-500 13271-502 13271-504 89400-956 89400-960
VWR® Flow-Through and Immersion Coolers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Enhance and simplify your low-temperature laboratory work with these low temperature coolers.
  • Compact add-on cooling
  • VWR® Two-Year Limited Parts and Labor Warranty
Available as immersion probe and flow-through models, these units provide rapid, low-cost cooling of liquids to temperatures as low as -100°C
Flow-through Style Coolers offer an extremely economical alternative to tap water cooling. Use with heated circulating baths to reach temperatures below ambient. Use with refrigerated circulating baths to increase cooling capacity.
Order Now
SPECIFICATIONS
Model Model 1107 Immersion Cooler, 1.75" Rigid Coil Model 1108 Flow-Through Cooler Model 1109 Immersion Probe Cooler, 3" Rigid Coil Immersion Probe Cooler, 3" Rigid Coil Immersion Probe Cooler, 1.5" Rigid Coil
Environmentally Preferable
Cooling capacity 106 W at -30°C,
152 W at -20°C,
253 W at -10°C,
421 W at 0°C,
703 W at +10°C,
1004 W at +20°C,
1205 W at +30°C,
1326 W at +40°C
117 W at -20°C,
260 W at -10°C,
322 W at 0°C,
537 W at +10°C,
745 W at +20°C,
921 W at +30°C,
1012 W at +40°C
117 W at -30°C,
164 W at -20°C,
273 W at -10°C,
454 W at 0°C,
757 W at +10°C,
1081 W at +20°C,
1298 W at +30°C,
1427 W at +40°C
0 W at -100°C,
35 W at -85°C,
85 W at -65°C
0 W at -65°C,
100 W at -40°C,
125 W at -30°C,
150 W at -20°C
Electrical 120V, 60Hz 120V, 60Hz 120V, 60Hz 120V, 60Hz 120V, 60Hz