1-Bromo-4-phenylnaphthalene ≥98.0% (by GC)

Supplier: TCI America

B5105-1G B5105-5G
76218-814EA 67.94 CAD
76218-814 76218-816
1-Bromo-4-phenylnaphthalene ≥98.0% (by GC)
1-Bromo-4-phenylnaphthalene
Formula: C₁₆H₁₁Br
Boiling Pt: 153°C
MDL Number: MFCD17012346
CAS Number: 59951-65-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR