2-Bromoallyl alcohol ≥97.0% (by GC)

Supplier: TCI America
Warning

B5347-1G B5347-5G
TCB5347-1GEA 161.16 CAD
TCB5347-1G TCB5347-5G
2-Bromoallyl alcohol ≥97.0% (by GC)
2-Bromoallyl alcohol
Formula: C₃H₅BrO
Boiling Pt: 150°C
Flash Pt: 76°C
MDL Number: MFCD09265077
CAS Number: 598-19-6

Order Now

Specification Test Results

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR