9,10-Diphenylanthracene-2-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
D4647-1G
D4647-200MG
TCD4647-1GEA
456.15
CAD
TCD4647-1G
TCD4647-200MG
9,10-Diphenylanthracene-2-boronic acid (contains varying amounts of Anhydride)
9,10-Diphenylanthracene-2-boronic acid
Formula:
C₂₆H₁₉BO₂ |
MDL Number:
MFCD16621110 CAS Number: 597553-98-5 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 106.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...