Anthracene-2,6-diyl bis(trifluoromethanesulfonate) ≥98.0% (by GC)

Supplier: TCI America
Warning

A3127-1G A3127-200MG
TCA3127-1GEA 584.6 CAD
TCA3127-1G TCA3127-200MG
Anthracene-2,6-diyl bis(trifluoromethanesulfonate) ≥98.0% (by GC)
Anthracene-2,6-diyl bis(trifluoromethanesulfonate)
Formula: C₁₆H₈F₆O₆S₂
CAS Number: 594838-61-6

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR