3-Bromopropyl Acetate ≥95.0% (by GC)

Supplier: TCI America

B5650-1G B5650-5G
TCB5650-1GEA 94.8 CAD
TCB5650-1G TCB5650-5G
3-Bromopropyl Acetate ≥95.0% (by GC)
3-Bromopropyl Acetate
Formula: C₅H₉BrO₂
Boiling Pt: 163°C
Storage Temperature: Refrigerator
MDL Number: MFCD02258489
CAS Number: 592-33-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR