Laurocapram 96,0 GC_ASSAY_METHOD

Supplier: TCI America

Synonyms: N-Dodecyl-ε-caprolactam, Azone, Laurocapram, N-Dodecyl-epsilon-caprolactam, 1-Dodecylazepan-2-one

L0317-25ML L0317-5ML
TCL0317-25MLEA 240.76 CAD
TCL0317-25ML TCL0317-5ML
Laurocapram 96,0 GC_ASSAY_METHOD
Laurocapram
Formula: C₁₈H₃₅NO
MW: 281.48 g/mol
Boiling Pt: 160°C
Melting Pt: -7°C
MDL Number: MFCD00190580
CAS Number: 59227-89-3
EINECS: 261-668-9
Merck Index: 14,05385

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR