2-Methoxyphenyl trifluoromethanesulfonate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2792-1G
M2792-5G
TCM2792-1GEA
75.84
CAD
TCM2792-1G
TCM2792-5G
2-Methoxyphenyl trifluoromethanesulfonate ≥98.0% (by GC)
2-Methoxyphenyl trifluoromethanesulfonate
Formula:
C₈H₇F₃O₄S Boiling Pt: 116°C |
MDL Number:
MFCD25541795 CAS Number: 59099-58-0 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...