2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3067-1G M3067-5G
TCM3067-1GEA 276.5 CAD
TCM3067-1G TCM3067-5G
2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)
2-Methoxy-N-(4-methoxyphenyl)aniline
Formula: C₁₄H₁₅NO₂
MDL Number: MFCD27956479
CAS Number: 58751-07-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR