Piperazine dihydriodide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

P2492-1G P2492-5G
TCP2492-1GEA 96.38 CAD
TCP2492-1G TCP2492-5G
Piperazine dihydriodide ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydriodide
Formula: C₄H₁₀N₂·₂HI
Melting Pt: 250°C
CAS Number: 58464-47-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR