6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one

C3138-1G C3138-5G
TCC3138-1GEA 176.96 CAD
TCC3138-1G TCC3138-5G
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by GC)
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Formula: C₉H₈ClNO₂
MW: 197.62 g/mol
MDL Number: MFCD01744501
CAS Number: 5791-00-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR