2-(4-Methoxyphenyl)indole ≥98.0% (by GC)

Supplier: TCI America

M2789-1G M2789-5G
76220-262EA 255.96 CAD
76220-262 76220-264
2-(4-Methoxyphenyl)indole ≥98.0% (by GC)
2-(4-Methoxyphenyl)indole
Formula: C₁₅H₁₃NO
MDL Number: MFCD00443483
CAS Number: 5784-95-2

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 229.0 to 233.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR