4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane ≥95.0% (by GC)

Supplier: TCI America
Warning

D5200-1G D5200-5G
TCD5200-1GEA 47.4 CAD
TCD5200-1G TCD5200-5G
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane ≥95.0% (by GC)
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 86°C
Storage Temperature: Refrigerator
MDL Number: MFCD16621162
CAS Number: 57280-22-5
EINECS: 421-750-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR