1,3-Bis(methoxymethoxy)benzene ≥98.0% (by GC)
Supplier: TCI America
B5643-1G
B5643-5G
76218-866EA
109.02
CAD
76218-866
76218-868
1,3-Bis(methoxymethoxy)benzene ≥98.0% (by GC)
1,3-Bis(methoxymethoxy)benzene
Formula:
C₁₀H₁₄O₄ Boiling Pt: 137°C |
MDL Number:
MFCD27925713 CAS Number: 57234-29-4 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...