4'-Cyano-4-dodecyloxybiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

C3270-1G C3270-5G
TCC3270-1GEA 289.14 CAD
TCC3270-1G TCC3270-5G
4'-Cyano-4-dodecyloxybiphenyl ≥98.0% (by GC)
4'-Cyano-4-dodecyloxybiphenyl
Formula: C₂₅H₃₃NO
MDL Number: MFCD01941019
CAS Number: 57125-50-5
EINECS: 260-575-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 84.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR