Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate ≥98.0% (by GC)
Supplier: TCI America
M2797-25G
M2797-5G
TCM2797-25GEA
919.56
CAD
TCM2797-25G
TCM2797-5G
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate ≥98.0% (by GC)
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate
Formula:
C₁₃H₁₆N₂O₆ |
MDL Number:
MFCD06797690 CAS Number: 57113-90-3 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...