9-(4-Iodophenyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

I0961-1G I0961-5G
TCI0961-1GEA 244.58 CAD
TCI0961-1G TCI0961-5G
9-(4-Iodophenyl)carbazole ≥98.0% (by GC)
9-(4-Iodophenyl)carbazole
Formula: C₁₈H₁₂IN
MDL Number: MFCD11617970
CAS Number: 57103-15-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR