1,2-Dimethyl-3H-benzo[e]indole ≥98.0% (by GC)

Supplier: TCI America

D5163-1G D5163-5G
76219-204EA 96.38 CAD
76219-204 76219-206
1,2-Dimethyl-3H-benzo[e]indole ≥98.0% (by GC)
1,2-Dimethyl-3H-benzo[e]indole
Formula: C₁₄H₁₃N
MDL Number: MFCD00850046
CAS Number: 55970-05-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR