S-(2-Methyl-3-furyl) Thioacetate ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Methyl-3-furanthiol Acetate, Thioacetic acid S-(2-Methyl-3-furyl) ester, S-(2-Methylfuran-3-yl) ethanethioate, 3-Acetylthio-2-methylfuran

M2661-25G M2661-5G
TCM2661-25GEA 165.9 CAD
TCM2661-25G TCM2661-5G
S-(2-Methyl-3-furyl) Thioacetate ≥96.0% (by GC)
S-(2-Methyl-3-furyl) Thioacetate
Formula: C₇H₈O₂S
Boiling Pt: 235°C
Flash Pt: 60°C
MDL Number: MFCD01632595
CAS Number: 55764-25-5
EINECS: 259-801-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR