Imidocarb Dipropionate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1,3-Bis(3-(4,5-dihydro-1H-imidazol-2-yl)phenyl)urea dipropionate

I0957-1G I0957-5G
TCI0957-1GEA 60.04 CAD
TCI0957-1G TCI0957-5G
Imidocarb Dipropionate ≥98.0% (by HPLC, titration analysis)
Imidocarb Dipropionate
Formula: C₁₉H₂₀N₆O·₂C₃H₆O₂
MW: 496.57 g/mol
MDL Number: MFCD00013161
CAS Number: 55750-06-6
EINECS: 259-791-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR