Hexadecamethylcyclooctasiloxane ≥96.0% (by GC)

Supplier: TCI America
Warning

H1288-1ML H1288-5ML
TCH1288-1MLEA 490.2 CAD
TCH1288-1ML TCH1288-5ML
Hexadecamethylcyclooctasiloxane ≥96.0% (by GC)
Hexadecamethylcyclooctasiloxane
Formula: C₁₆H₄₈O₈Si₈
Boiling Pt: 168°C
CAS Number: 556-68-3
EINECS: 209-137-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR