(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Ikeda - Omote Reagent

T2977-1G T2977-5G
TCT2977-1GEA 1564.2 CAD
TCT2977-1G TCT2977-5G
(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane ≥95.0% (by GC)
(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane
Formula: C₆H₁₁F₃Si
Boiling Pt: 69°C
CAS Number: 55364-28-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR