N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide ≥92.0% (by total nitrogen basis)

Supplier: TCI America
Warning

D4429-1G D4429-5G
TCD4429-1GEA 218.04 CAD
TCD4429-1G TCD4429-5G
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide ≥92.0% (by total nitrogen basis)
N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic diimide
Formula: C₂₆H₁₄N₂O₄
MDL Number: MFCD00071975
CAS Number: 5521-31-3
EINECS: 226-866-1

Order Now

Specification Test Results

Appearance Orange to Brown to Dark purple powder to crystal
Purity (with Total Nitrogen) min. 92.0 %
Lambda max. 590.0 to 600.0 nm (H2SO4)
Molar extinction coefficient min.76000 (H2SO4, 590.0 to 600.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR