Tris(4-aminophenyl)methane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
T3599-1G
T3599-5G
TCT3599-1GEA
268.41
CAD
TCT3599-1G
TCT3599-5G
Tris(4-aminophenyl)methane ≥97.0% (by GC, titration analysis)
Tris(4-aminophenyl)methane
Formula:
C₁₉H₁₉N₃ Melting Pt: 208°C |
MDL Number:
MFCD00137820 CAS Number: 548-61-8 EINECS: 208-952-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...