1,4-Diazabicyclo[2.2.2]octane dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
D5250-1G
D5250-5G
76219-876EA
161.16
CAD
76219-876
76219-878
1,4-Diazabicyclo[2.2.2]octane dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
1,4-Diazabicyclo[2.2.2]octane dihydrobromide
Formula:
C₆H₁₂N₂·₂HBr Melting Pt: 319°C |
MDL Number:
MFCD30725750 CAS Number: 54581-69-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...