Aniline hydrobromide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

A2985-1G A2985-5G
TCA2985-1GEA 94.89 CAD
TCA2985-1G TCA2985-5G
Aniline hydrobromide ≥98.0% (by HPLC)
Aniline hydrobromide
Formula: C₆H₇N·HBr
Melting Pt: 285°C
MDL Number: MFCD00035465
CAS Number: 542-11-0
EINECS: 208-801-9

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR