(R)-(+)-2-Acetoxypropionyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
A3085-1G
A3085-5G
76218-794EA
213.3
CAD
76218-794
76218-796
(R)-(+)-2-Acetoxypropionyl chloride ≥98.0% (by titrimetric analysis)
(R)-(+)-2-Acetoxypropionyl chloride
Formula:
C₅H₇ClO₃ Boiling Pt: 53°C Storage Temperature: Refrigerator |
MDL Number:
MFCD27925055 CAS Number: 53636-19-4 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Optical purity (GC) | min. 96.0 ee% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...