1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane ≥98.0% (by GC)

Supplier: TCI America
Warning

H1613-25ML H1613-5ML
76220-158EA 237 CAD
76220-158 76220-160
1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane ≥98.0% (by GC)
1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane
Formula: C₁₁H₃₀O₃Si₄
Boiling Pt: 88°C
Flash Pt: 66°C
MDL Number: MFCD00054159
CAS Number: 5356-84-3
EINECS: 226-342-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR