5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Dibenzo[a,e]cyclooctadien-5,11-diyne, 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene

T3241-100MG T3241-500MG
TCT3241-100MGEA 382.36 CAD
TCT3241-100MG TCT3241-500MG
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene ≥98.0% (by HPLC)
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene
Formula: C₁₆H₈
MW: 200.24 g/mol
Melting Pt: 110°C
Storage Temperature: Freezer
MDL Number: MFCD28386121
CAS Number: 53397-65-2

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR