7-(Bromomethyl)pentadecane ≥95.0% (by GC)

Supplier: TCI America
Warning

B4544-25ML B4544-5ML
TCB4544-25MLEA 188.49 CAD
TCB4544-25ML TCB4544-5ML
7-(Bromomethyl)pentadecane ≥95.0% (by GC)
7-(Bromomethyl)pentadecane
Formula: C₁₆H₃₃Br
Boiling Pt: 118°C
Flash Pt: 108°C
MDL Number: MFCD00136231
CAS Number: 52997-43-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR