N,4-Dimethoxy-N-methylbenzamide ≥95.0% (by GC)

Supplier: TCI America
Warning

D5475-1G D5475-5G
TCD5475-1GEA 66.36 CAD
TCD5475-1G TCD5475-5G
N,4-Dimethoxy-N-methylbenzamide ≥95.0% (by GC)
N,4-Dimethoxy-N-methylbenzamide
Formula: C₁₀H₁₃NO₃
Boiling Pt: 105°C
MDL Number: MFCD02684314
CAS Number: 52898-49-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR