Kirenol ≥98.0% (by GC)

Supplier: TCI America

K0059-100MG K0059-25MG
TCK0059-100MGEA 891.12 CAD
TCK0059-100MG TCK0059-25MG
Kirenol ≥98.0% (by GC)
Kirenol
Formula: C₂₀H₃₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00210519
CAS Number: 52659-56-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C
Specific rotation (value) -32.0 to -29.0 deg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR