3-Chloro-4-(2-pyridylmethoxy)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3150-1G C3150-5G
TCC3150-1GEA 218.04 CAD
TCC3150-1G TCC3150-5G
3-Chloro-4-(2-pyridylmethoxy)aniline ≥98.0% (by GC, titration analysis)
3-Chloro-4-(2-pyridylmethoxy)aniline
Formula: C₁₂H₁₁ClN₂O
MDL Number: MFCD08699389
CAS Number: 524955-09-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR